The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032108.3 | NP_115484.2 | 2 - 3 | 379 | 59 |
XM_011527639.2 | XP_011525941.1 | 2 - 3 | 428 | 59 | |
XM_011527640.2 | XP_011525942.1 | 2 - 3 | 171 | 59 | |
XM_011527641.1 | XP_011525943.1 | 2 - 3 | 428 | 59 | |
GenBank mRNA | AB022433.1 | - | 1 - 2 | 155 | 59 |
AB209278.1 | - | 1 - 2 | 141 | 59 | |
AF216389.1 | - | 1 - 2 | 120 | 59 | |
AF293363.1 | - | 1 - 2 | 120 | 59 | |
AK300037.1 | - | 2 - 3 | 379 | 59 | |
AY358939.1 | - | 1 - 2 | 135 | 59 | |
BC142617.1 | - | 2 - 3 | 322 | 59 | |
DA818632.1 | - | 2 - 3 | 379 | 59 | |
DB257858.1 | - | 2 - 3 | 429 | 59 | |
FR839673.1 | - | 1 - 2 | 117 | 59 |