Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE4
Entrez Gene ID
163183
Gene Name
spectrin repeat containing nuclear envelope family member 4
Gene Aliases
C19orf46, DFNB76, Nesp4
Chromosome Location
Chr.19: 36003299 - 36008793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039876.2 NP_001034965.1 2 - 3 390 143
GenBank mRNA AK093764.1 - 2 - 3 364 143

View Full Product Details