The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242783.1 | NP_001229712.1 | 5 - 6 | 1110 | 91 |
NM_003449.4 | NP_003440.1 | 6 - 7 | 1331 | 91 | |
XM_005249374.2 | XP_005249431.1 | 6 - 7 | 1127 | 91 | |
XM_005249375.2 | XP_005249432.1 | 6 - 7 | 1135 | 91 | |
XM_005249376.2 | XP_005249433.1 | 6 - 7 | 1207 | 91 | |
XM_005249377.2 | XP_005249434.1 | 5 - 6 | 967 | 91 | |
XM_005249378.2 | XP_005249435.1 | 4 - 5 | 947 | 91 | |
XM_006715180.2 | XP_006715243.1 | 5 - 6 | 1151 | 91 | |
GenBank mRNA | AK314782.1 | - | 6 - 7 | 1093 | 91 |
BC024039.1 | - | 6 - 7 | 1185 | 91 | |
BC032297.1 | - | 5 - 6 | 968 | 91 |