The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002841.3 | NP_002832.3 | 9 - 10 | 1940 | 71 |
XM_005265353.3 | XP_005265410.1 | 9 - 10 | 1598 | 71 | |
XM_017006961.1 | XP_016862450.1 | 10 - 11 | 1456 | 71 | |
XM_017006962.1 | XP_016862451.1 | 10 - 11 | 1275 | 71 | |
XM_017006963.1 | XP_016862452.1 | 10 - 11 | 1456 | 71 | |
XM_017006964.1 | XP_016862453.1 | 9 - 10 | 2326 | 71 | |
GenBank mRNA | AB209871.1 | - | 9 - 10 | 1586 | 71 |
BC140904.1 | - | 9 - 10 | 1598 | 71 | |
BC144110.1 | - | 9 - 10 | 1598 | 71 | |
L09247.1 | - | 9 - 10 | 1300 | 71 |