The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145277.1 | NP_001138749.1 | 4 - 5 | 471 | 73 |
NM_001145278.1 | NP_001138750.1 | 4 - 5 | 393 | 73 | |
NM_018090.4 | NP_060560.1 | 4 - 5 | 471 | 73 | |
XM_011541767.2 | XP_011540069.1 | 5 - 6 | 543 | 73 | |
XM_017001721.1 | XP_016857210.1 | 4 - 5 | 419 | 73 | |
GenBank mRNA | AK001282.1 | - | 4 - 5 | 403 | 73 |
AK023545.1 | - | 4 - 5 | 407 | 73 | |
AK024468.1 | - | 4 - 5 | 385 | 73 | |
AK223344.1 | - | 4 - 5 | 419 | 73 | |
AK300764.1 | - | 4 - 5 | 417 | 73 | |
AK302227.1 | - | 4 - 5 | 339 | 73 | |
BC017014.1 | - | 4 - 5 | 402 | 73 | |
BC018914.2 | - | 4 - 5 | 405 | 73 |