The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035.2 | NP_001026.2 | 22 - 23 | 2731 | 136 |
XM_006711802.3 | XP_006711865.1 | 22 - 23 | 2925 | 136 | |
XM_006711803.3 | XP_006711866.1 | 22 - 23 | 2925 | 136 | |
XM_006711804.3 | XP_006711867.1 | 22 - 23 | 2925 | 136 | |
XM_006711805.3 | XP_006711868.1 | 22 - 23 | 2925 | 136 | |
XM_006711806.3 | XP_006711869.1 | 22 - 23 | 2925 | 136 | |
XM_006711807.3 | XP_006711870.1 | 22 - 23 | 2925 | 136 | |
XM_006711808.3 | XP_006711871.1 | 22 - 23 | 2925 | 136 | |
XM_006711809.3 | XP_006711872.1 | 22 - 23 | 2925 | 136 | |
XM_006711810.3 | XP_006711873.1 | 22 - 23 | 2925 | 136 | |
XM_017002028.1 | XP_016857517.1 | 21 - 22 | 2904 | 136 | |
GenBank mRNA | X98330.1 | - | 22 - 23 | 2732 | 136 |