The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035.2 | NP_001026.2 | 30 - 31 | 3924 | 78 |
XM_006711802.3 | XP_006711865.1 | 30 - 31 | 4118 | 78 | |
XM_006711803.3 | XP_006711866.1 | 30 - 31 | 4115 | 78 | |
XM_006711804.3 | XP_006711867.1 | 30 - 31 | 4118 | 78 | |
XM_006711805.3 | XP_006711868.1 | 30 - 31 | 4118 | 78 | |
XM_006711806.3 | XP_006711869.1 | 30 - 31 | 4118 | 78 | |
XM_006711807.3 | XP_006711870.1 | 30 - 31 | 4118 | 78 | |
XM_006711808.3 | XP_006711871.1 | 30 - 31 | 4118 | 78 | |
XM_006711809.3 | XP_006711872.1 | 30 - 31 | 4118 | 78 | |
XM_006711810.3 | XP_006711873.1 | 30 - 31 | 4115 | 78 | |
XM_017002028.1 | XP_016857517.1 | 29 - 30 | 4097 | 78 | |
GenBank mRNA | X98330.1 | - | 30 - 31 | 3925 | 78 |