Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RYR2
Entrez Gene ID
6262
Gene Name
ryanodine receptor 2
Gene Aliases
ARVC2, ARVD2, RYR-2, RyR, VTSIP
Chromosome Location
Chr.1: 237042208 - 237833988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035.2 NP_001026.2 84 - 85 11593 63
XM_006711802.3 XP_006711865.1 86 - 87 11841 63
XM_006711803.3 XP_006711866.1 86 - 87 11838 63
XM_006711804.3 XP_006711867.1 85 - 86 11817 63
XM_006711805.3 XP_006711868.1 85 - 86 11811 63
XM_006711806.3 XP_006711869.1 85 - 86 11805 63
XM_006711807.3 XP_006711870.1 84 - 85 11781 63
XM_006711808.3 XP_006711871.1 83 - 84 11604 63
XM_006711810.3 XP_006711873.1 83 - 84 11748 63
XM_017002028.1 XP_016857517.1 85 - 86 11820 63
GenBank mRNA AK294792.1 - 24 - 25 2668 63
X98330.1 - 84 - 85 11594 63

View Full Product Details