Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTPRF
Entrez Gene ID
5792
Gene Name
protein tyrosine phosphatase, receptor type F
Gene Aliases
BNAH2, LAR
Chromosome Location
Chr.1: 43522238 - 43623672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005271079.3 XP_005271136.1 26 - 27 4598 82
XM_005271080.3 XP_005271137.1 25 - 26 4586 82
XM_005271081.3 XP_005271138.1 24 - 25 4559 82
XM_005271082.3 XP_005271139.1 22 - 23 4220 82
XM_006710795.3 XP_006710858.1 27 - 28 4576 82
XM_006710796.3 XP_006710859.1 25 - 26 4571 82
XM_006710797.3 XP_006710860.1 25 - 26 4565 82
XM_006710798.3 XP_006710861.1 24 - 25 4538 82
XM_006710799.3 XP_006710862.1 25 - 26 4292 82
XM_006710800.3 XP_006710863.1 24 - 25 4265 82
XM_006710801.3 XP_006710864.1 25 - 26 4019 82
XM_011541871.2 XP_011540173.1 27 - 28 4610 82
XM_011541872.2 XP_011540174.1 27 - 28 4596 82
XM_011541873.2 XP_011540175.1 27 - 28 4530 82
XM_017001942.1 XP_016857431.1 24 - 25 4227 82
XM_017001943.1 XP_016857432.1 24 - 25 4280 82
XM_017001944.1 XP_016857433.1 23 - 24 4232 82
XM_017001945.1 XP_016857434.1 23 - 24 4253 82
XM_017001946.1 XP_016857435.1 24 - 25 3713 82
XM_017001947.1 XP_016857436.1 23 - 24 3686 82
XM_017001948.1 XP_016857437.1 23 - 24 3712 82
XM_017001949.1 XP_016857438.1 24 - 25 3677 82
XM_017001950.1 XP_016857439.1 21 - 22 3649 82
GenBank mRNA AB177856.1 - 24 - 25 3210 82
AB177857.1 - 23 - 24 4485 82
AB209478.1 - 15 - 16 3134 82
AK125325.1 - 15 - 16 2470 82
AK127007.1 - 12 - 13 1413 82
AK127475.1 - 2 - 3 702 82
BC048768.1 - 23 - 24 4496 82
BX537361.1 - 11 - 12 2697 82
Y00815.1 - 24 - 25 4537 82

View Full Product Details