Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTPRF
Entrez Gene ID
5792
Gene Name
protein tyrosine phosphatase, receptor type F
Gene Aliases
BNAH2, LAR
Chromosome Location
Chr.1: 43522238 - 43623672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005271079.3 XP_005271136.1 34 - 35 5922 77
XM_005271080.3 XP_005271137.1 33 - 34 5910 77
XM_005271081.3 XP_005271138.1 32 - 33 5883 77
XM_005271082.3 XP_005271139.1 30 - 31 5544 77
XM_006710795.3 XP_006710858.1 35 - 36 5900 77
XM_006710796.3 XP_006710859.1 33 - 34 5895 77
XM_006710797.3 XP_006710860.1 33 - 34 5889 77
XM_006710798.3 XP_006710861.1 32 - 33 5862 77
XM_006710799.3 XP_006710862.1 33 - 34 5616 77
XM_006710800.3 XP_006710863.1 32 - 33 5589 77
XM_006710801.3 XP_006710864.1 33 - 34 5343 77
XM_011541871.2 XP_011540173.1 35 - 36 5934 77
XM_011541872.2 XP_011540174.1 35 - 36 5920 77
XM_011541873.2 XP_011540175.1 35 - 36 5854 77
XM_017001942.1 XP_016857431.1 32 - 33 5551 77
XM_017001943.1 XP_016857432.1 32 - 33 5604 77
XM_017001944.1 XP_016857433.1 31 - 32 5556 77
XM_017001945.1 XP_016857434.1 31 - 32 5577 77
XM_017001946.1 XP_016857435.1 32 - 33 5037 77
XM_017001947.1 XP_016857436.1 31 - 32 5010 77
XM_017001948.1 XP_016857437.1 31 - 32 5036 77
XM_017001949.1 XP_016857438.1 32 - 33 5001 77
XM_017001950.1 XP_016857439.1 29 - 30 4973 77
GenBank mRNA AB177856.1 - 32 - 33 4534 77
AB177857.1 - 31 - 32 5809 77
AB209478.1 - 23 - 24 4458 77
AK125325.1 - 23 - 24 3794 77
AK127007.1 - 20 - 21 2737 77
AK127475.1 - 9 - 10 2258 77
AK301551.1 - 6 - 7 892 77
BC048768.1 - 31 - 32 5820 77
BX537361.1 - 19 - 20 4021 77
Y00815.1 - 32 - 33 5861 77

View Full Product Details