Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRF
Entrez Gene ID
5792
Gene Name
protein tyrosine phosphatase, receptor type F
Gene Aliases
BNAH2, LAR
Chromosome Location
Chr.1: 43522238 - 43623672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005271079.3 XP_005271136.1 35 - 36 6051 71
XM_005271080.3 XP_005271137.1 34 - 35 6039 71
XM_005271081.3 XP_005271138.1 33 - 34 6012 71
XM_005271082.3 XP_005271139.1 31 - 32 5673 71
XM_006710795.3 XP_006710858.1 36 - 37 6029 71
XM_006710796.3 XP_006710859.1 34 - 35 6024 71
XM_006710797.3 XP_006710860.1 34 - 35 6018 71
XM_006710798.3 XP_006710861.1 33 - 34 5991 71
XM_006710799.3 XP_006710862.1 34 - 35 5745 71
XM_006710800.3 XP_006710863.1 33 - 34 5718 71
XM_006710801.3 XP_006710864.1 34 - 35 5472 71
XM_011541871.2 XP_011540173.1 36 - 37 6063 71
XM_011541872.2 XP_011540174.1 36 - 37 6049 71
XM_011541873.2 XP_011540175.1 36 - 37 5983 71
XM_017001942.1 XP_016857431.1 33 - 34 5680 71
XM_017001943.1 XP_016857432.1 33 - 34 5733 71
XM_017001944.1 XP_016857433.1 32 - 33 5685 71
XM_017001945.1 XP_016857434.1 32 - 33 5706 71
XM_017001946.1 XP_016857435.1 33 - 34 5166 71
XM_017001947.1 XP_016857436.1 32 - 33 5139 71
XM_017001948.1 XP_016857437.1 32 - 33 5165 71
XM_017001949.1 XP_016857438.1 33 - 34 5130 71
XM_017001950.1 XP_016857439.1 30 - 31 5102 71
GenBank mRNA AB177856.1 - 33 - 34 4663 71
AB177857.1 - 32 - 33 5938 71
AB209478.1 - 24 - 25 4587 71
AK125325.1 - 24 - 25 3923 71
AK127007.1 - 21 - 22 2866 71
AK127475.1 - 10 - 11 2387 71
AK301551.1 - 7 - 8 1021 71
BC048768.1 - 32 - 33 5949 71
BX537361.1 - 20 - 21 71
Y00815.1 - 33 - 34 5990 71

View Full Product Details