The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287811.1 | NP_001274740.1 | 3 - 4 | 554 | 66 |
NM_015291.3 | NP_056106.1 | 4 - 5 | 739 | 66 | |
NR_109898.1 | 4 - 5 | 739 | 66 | ||
GenBank mRNA | AB023179.2 | - | 4 - 5 | 695 | 66 |
AK091848.1 | - | 4 - 5 | 697 | 66 | |
AK292479.1 | - | 4 - 5 | 671 | 66 | |
AL833299.1 | - | 4 - 5 | 678 | 66 | |
BC047363.1 | - | 4 - 5 | 739 | 66 | |
BX648982.1 | - | 4 - 5 | 664 | 66 | |
CR749259.1 | - | 3 - 4 | 499 | 66 | |
CR749567.1 | - | 3 - 4 | 499 | 66 |