Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
Chromosome Location
Chr.22: 32413845 - 32464465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174932.2 NP_777592.1 13 - 14 1322 164
XM_011530088.2 XP_011528390.1 15 - 16 1502 164
XM_011530089.1 XP_011528391.1 15 - 16 1459 164
XM_011530090.1 XP_011528392.1 14 - 15 1360 164
XM_011530091.2 XP_011528393.1 14 - 15 1346 164
XM_017028740.1 XP_016884229.1 14 - 15 1331 164
GenBank mRNA AF465766.1 - 13 - 14 1322 164
AK301685.1 - 13 - 14 1228 164
BC131582.1 - 13 - 14 1237 164
CU456409.1 - 4 - 5 370 164

View Full Product Details