The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164239.1 | NP_001157711.1 | 15 - 16 | 2316 | 58 |
NM_003587.4 | NP_003578.2 | 15 - 16 | 2637 | 58 | |
XM_011514938.2 | XP_011513240.1 | 12 - 13 | 2430 | 58 | |
XM_011514939.2 | XP_011513241.1 | 12 - 13 | 1904 | 58 | |
XM_011514940.2 | XP_011513242.1 | 15 - 16 | 2845 | 58 | |
XM_011514941.2 | XP_011513243.1 | 7 - 8 | 929 | 58 | |
GenBank mRNA | AB001601.1 | - | 15 - 16 | 2564 | 58 |
AF119860.1 | - | 7 - 8 | 1109 | 58 | |
AK302721.1 | - | 15 - 16 | 2310 | 58 | |
AK312264.1 | - | 15 - 16 | 2576 | 58 | |
BC008825.2 | - | 15 - 16 | 2588 | 58 | |
BC009392.2 | - | 15 - 16 | 2588 | 58 |