The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164239.1 | NP_001157711.1 | 16 - 17 | 2396 | 70 |
NM_003587.4 | NP_003578.2 | 16 - 17 | 2717 | 70 | |
XM_011514938.2 | XP_011513240.1 | 13 - 14 | 2510 | 70 | |
XM_011514939.2 | XP_011513241.1 | 13 - 14 | 1984 | 70 | |
XM_011514940.2 | XP_011513242.1 | 16 - 17 | 2925 | 70 | |
XM_011514941.2 | XP_011513243.1 | 8 - 9 | 1009 | 70 | |
GenBank mRNA | AB001601.1 | - | 16 - 17 | 2644 | 70 |
AF119860.1 | - | 8 - 9 | 1189 | 70 | |
AK302721.1 | - | 16 - 17 | 2390 | 70 | |
AK312264.1 | - | 16 - 17 | 2656 | 70 | |
BC008825.2 | - | 16 - 17 | 2668 | 70 | |
BC009392.2 | - | 16 - 17 | 2668 | 70 |