The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164239.1 | NP_001157711.1 | 18 - 19 | 2719 | 88 |
NM_003587.4 | NP_003578.2 | 18 - 19 | 3040 | 88 | |
XM_011514938.2 | XP_011513240.1 | 15 - 16 | 2833 | 88 | |
XM_011514939.2 | XP_011513241.1 | 15 - 16 | 2307 | 88 | |
XM_011514940.2 | XP_011513242.1 | 18 - 19 | 3248 | 88 | |
XM_011514941.2 | XP_011513243.1 | 10 - 11 | 1332 | 88 | |
GenBank mRNA | AB001601.1 | - | 18 - 19 | 2967 | 88 |
AF119860.1 | - | 10 - 11 | 1512 | 88 | |
AK312264.1 | - | 18 - 19 | 2979 | 88 | |
BC008825.2 | - | 18 - 19 | 2991 | 88 | |
BC009392.2 | - | 18 - 19 | 2991 | 88 | |
BQ772493.1 | - | 2 - 3 | 88 |