Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP42
Entrez Gene ID
143872
Gene Name
Rho GTPase activating protein 42
Gene Aliases
GRAF3
Chromosome Location
Chr.11: 100687676 - 100993941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011542615.2 XP_011540917.1 24 - 24 9883 94
XM_011542616.2 XP_011540918.1 24 - 24 7652 94
XM_011542617.2 XP_011540919.1 25 - 25 7538 94
XM_017017237.1 XP_016872726.1 24 - 24 7841 94
XM_017017238.1 XP_016872727.1 25 - 25 10024 94

Gene Symbol
TMEM133
Entrez Gene ID
83935
Gene Name
transmembrane protein 133
Gene Aliases
AD031
Chromosome Location
Chr.11: 100992080 - 100993935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032021.2 NP_114410.1 1 - 1 1489 94
GenBank mRNA AF247167.1 - 1 - 1 1500 94

View Full Product Details