The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288750.1 | NP_001275679.1 | 6 - 7 | 671 | 104 |
NM_001288751.1 | NP_001275680.1 | 7 - 8 | 878 | 104 | |
NM_030770.3 | NP_110397.2 | 7 - 8 | 774 | 104 | |
NR_110046.1 | 5 - 6 | 572 | 104 | ||
NR_110047.1 | 5 - 6 | 572 | 104 | ||
XM_017018366.1 | XP_016873855.1 | 5 - 6 | 4010 | 104 | |
XM_017018367.1 | XP_016873856.1 | 7 - 8 | 4202 | 104 | |
GenBank mRNA | AB028140.1 | - | 7 - 8 | 780 | 104 |
AL538140.3 | - | 7 - 8 | 715 | 104 | |
BC121802.1 | - | 6 - 7 | 644 | 104 | |
BC121803.1 | - | 7 - 8 | 851 | 104 |