Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS5
Entrez Gene ID
80975
Gene Name
transmembrane protease, serine 5
Gene Aliases
SPINESIN
Chromosome Location
Chr.11: 113687546 - 113709811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288749.1 NP_001275678.1 6 - 7 799 116
NM_001288750.1 NP_001275679.1 8 - 9 1006 116
NM_001288751.1 NP_001275680.1 9 - 10 1213 116
NM_001288752.1 NP_001275681.1 7 - 8 902 116
NM_030770.3 NP_110397.2 9 - 10 1109 116
NR_110046.1 7 - 8 907 116
NR_110047.1 7 - 8 907 116
XM_017018366.1 XP_016873855.1 6 - 7 5573 116
XM_017018367.1 XP_016873856.1 8 - 9 5765 116
GenBank mRNA AB028140.1 - 9 - 10 1115 116
AK294313.1 - 6 - 7 734 116
AK295366.1 - 7 - 8 831 116
BC121802.1 - 8 - 9 979 116
BC121803.1 - 9 - 10 1186 116

View Full Product Details