The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138800.1 | NP_620155.1 | 2 - 3 | 554 | 120 |
GenBank mRNA | BC015353.1 | - | 2 - 3 | 554 | 120 |