Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SETD5
Entrez Gene ID
55209
Gene Name
SET domain containing 5
Gene Aliases
-
Chromosome Location
Chr.3: 9397700 - 9478154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080517.2 NP_001073986.1 19 - 20 3651 63
NM_001292043.1 NP_001278972.1 21 - 22 3915 63
XM_005265301.1 XP_005265358.1 19 - 20 3689 63
XM_005265303.1 XP_005265360.1 22 - 23 4127 63
XM_011533920.1 XP_011532222.1 23 - 24 4222 63
XM_011533921.1 XP_011532223.1 22 - 23 3824 63
XM_011533922.1 XP_011532224.1 21 - 22 3785 63
XM_011533927.1 XP_011532229.1 23 - 24 4165 63
XM_011533928.1 XP_011532230.1 21 - 22 3728 63
XM_011533930.1 XP_011532232.1 23 - 24 4166 63
XM_011533932.1 XP_011532234.1 21 - 22 3933 63
XM_011533933.1 XP_011532235.1 19 - 20 3302 63
XM_017006767.1 XP_016862256.1 23 - 24 5884 63
XM_017006768.1 XP_016862257.1 21 - 22 5398 63
XM_017006769.1 XP_016862258.1 21 - 22 7501 63
XM_017006770.1 XP_016862259.1 21 - 22 7483 63
XM_017006771.1 XP_016862260.1 20 - 21 3746 63
XM_017006772.1 XP_016862261.1 20 - 21 7444 63
XM_017006773.1 XP_016862262.1 21 - 22 7444 63
XM_017006774.1 XP_016862263.1 20 - 21 3689 63
XM_017006775.1 XP_016862264.1 20 - 21 7387 63
XM_017006776.1 XP_016862265.1 22 - 23 4029 63
XM_017006777.1 XP_016862266.1 22 - 23 3972 63
XM_017006778.1 XP_016862267.1 23 - 24 4013 63
XM_017006779.1 XP_016862268.1 21 - 22 3990 63
XM_017006780.1 XP_016862269.1 22 - 23 3974 63
XM_017006781.1 XP_016862270.1 22 - 23 3956 63
XM_017006782.1 XP_016862271.1 22 - 23 7540 63
XM_017006783.1 XP_016862272.1 24 - 25 4238 63
GenBank mRNA AB051544.1 - 15 - 16 3445 63
AK001569.1 - 8 - 9 1834 63
AK024384.1 - 7 - 8 1745 63
AK025478.1 - 3 - 4 759 63
AK091393.1 - 16 - 17 2954 63
AK127298.1 - 9 - 10 2538 63
AK289529.1 - 10 - 11 2233 63
AL442073.1 - 6 - 7 1631 63
BC020956.1 - 8 - 9 1865 63
BX648380.1 - 11 - 12 2253 63
CR627408.1 - 18 - 19 3267 63

View Full Product Details