Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SETD5
Entrez Gene ID
55209
Gene Name
SET domain containing 5
Gene Aliases
-
Chromosome Location
Chr.3: 9397700 - 9478154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080517.2 NP_001073986.1 9 - 10 1404 96
NM_001292043.1 NP_001278972.1 11 - 12 1668 96
XM_005265301.1 XP_005265358.1 9 - 10 1442 96
XM_005265303.1 XP_005265360.1 11 - 12 1823 96
XM_011533920.1 XP_011532222.1 12 - 13 1918 96
XM_011533921.1 XP_011532223.1 11 - 12 1520 96
XM_011533922.1 XP_011532224.1 10 - 11 1481 96
XM_011533927.1 XP_011532229.1 12 - 13 1861 96
XM_011533928.1 XP_011532230.1 10 - 11 1424 96
XM_011533930.1 XP_011532232.1 12 - 13 1862 96
XM_011533932.1 XP_011532234.1 10 - 11 1629 96
XM_011533933.1 XP_011532235.1 8 - 9 998 96
XM_017006767.1 XP_016862256.1 12 - 13 3580 96
XM_017006768.1 XP_016862257.1 10 - 11 3094 96
XM_017006769.1 XP_016862258.1 10 - 11 5197 96
XM_017006770.1 XP_016862259.1 11 - 12 5236 96
XM_017006771.1 XP_016862260.1 9 - 10 1442 96
XM_017006772.1 XP_016862261.1 10 - 11 5197 96
XM_017006773.1 XP_016862262.1 10 - 11 5140 96
XM_017006774.1 XP_016862263.1 9 - 10 1385 96
XM_017006775.1 XP_016862264.1 10 - 11 5140 96
XM_017006776.1 XP_016862265.1 11 - 12 1725 96
XM_017006777.1 XP_016862266.1 11 - 12 1668 96
XM_017006778.1 XP_016862267.1 12 - 13 1709 96
XM_017006779.1 XP_016862268.1 10 - 11 1686 96
XM_017006780.1 XP_016862269.1 11 - 12 1670 96
XM_017006781.1 XP_016862270.1 12 - 13 1709 96
XM_017006782.1 XP_016862271.1 11 - 12 5236 96
XM_017006783.1 XP_016862272.1 13 - 14 1934 96
XM_017006784.1 XP_016862273.1 11 - 12 5236 96
XM_017006785.1 XP_016862274.1 11 - 12 5236 96
XM_017006786.1 XP_016862275.1 11 - 12 5236 96
GenBank mRNA AB051544.1 - 5 - 6 1198 96
AK091393.1 - 6 - 7 707 96
AK092046.1 - 11 - 12 1623 96
CR627408.1 - 8 - 9 1020 96

View Full Product Details