The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000393.3 | NP_000384.2 | 24 - 25 | 1890 | 61 |
XM_011510573.2 | XP_011508875.1 | 24 - 25 | 1672 | 61 | |
GenBank mRNA | AB209045.1 | - | 24 - 25 | 1752 | 61 |
AK297936.1 | - | 24 - 25 | 1772 | 61 | |
BC086874.1 | - | 8 - 9 | 879 | 61 | |
X04758.1 | - | 6 - 7 | 415 | 61 | |
Y14690.1 | - | 24 - 25 | 1744 | 61 |