Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TP53INP2
Entrez Gene ID
58476
Gene Name
tumor protein p53 inducible nuclear protein 2
Gene Aliases
C20orf110, DOR, PIG-U, PINH, dJ1181N3.1
Chromosome Location
Chr.20: 34704303 - 34713439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011528952.2 XP_011527254.1 2 - 3 327 91
GenBank mRNA AJ297792.1 - 2 - 3 274 91
AK292443.1 - 2 - 3 274 91
AY422170.1 - 2 - 3 283 91
BC035639.1 - 2 - 3 172 91
HY108580.1 - 2 - 3 322 91
HY157843.1 - 2 - 3 152 91

View Full Product Details