The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018024.2 | NP_060494.1 | 1 - 2 | 205 | 153 |
XM_006716597.3 | XP_006716660.1 | 1 - 2 | 199 | 153 | |
XM_011517147.1 | XP_011515449.1 | 1 - 2 | 198 | 153 | |
XM_011517151.2 | XP_011515453.1 | 1 - 2 | 196 | 153 | |
XM_017013601.1 | XP_016869090.1 | 1 - 2 | 199 | 153 | |
GenBank mRNA | AK001066.1 | - | 1 - 2 | 205 | 153 |
AK225643.1 | - | 1 - 2 | 156 | 153 | |
AU119047.1 | - | 1 - 2 | 205 | 153 | |
BC008781.2 | - | 1 - 2 | 123 | 153 |