The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282944.1 | NP_001269873.1 | 10 - 11 | 1265 | 110 |
NM_001282946.1 | NP_001269875.1 | 11 - 12 | 1358 | 110 | |
NM_015160.2 | NP_055975.1 | 11 - 12 | 1360 | 110 | |
XM_005266059.4 | XP_005266116.1 | 11 - 12 | 1275 | 110 | |
XM_011518417.2 | XP_011516719.1 | 7 - 8 | 814 | 110 | |
XM_017014543.1 | XP_016870032.1 | 10 - 11 | 1221 | 110 | |
GenBank mRNA | AK296617.1 | - | 10 - 11 | 1177 | 110 |
AK308104.1 | - | 11 - 12 | 1273 | 110 | |
BC004342.1 | - | 2 - 3 | 119 | 110 | |
BC022949.1 | - | 11 - 12 | 1252 | 110 | |
BC033103.1 | - | 11 - 12 | 1269 | 110 | |
BC111399.1 | - | 11 - 12 | 1275 | 110 | |
BC132724.1 | - | 11 - 12 | 1299 | 110 | |
BC136599.1 | - | 11 - 12 | 1299 | 110 | |
D21064.1 | - | 11 - 12 | 1273 | 110 | |
D50913.2 | - | 11 - 12 | 1273 | 110 |