The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282944.1 | NP_001269873.1 | 2 - 3 | 363 | 134 |
NM_001282946.1 | NP_001269875.1 | 2 - 3 | 363 | 134 | |
NM_015160.2 | NP_055975.1 | 2 - 3 | 363 | 134 | |
XM_005266059.4 | XP_005266116.1 | 2 - 3 | 278 | 134 | |
XM_017014543.1 | XP_016870032.1 | 1 - 2 | 196 | 134 | |
GenBank mRNA | AK296617.1 | - | 2 - 3 | 275 | 134 |
AK299304.1 | - | 2 - 3 | 295 | 134 | |
AK308104.1 | - | 2 - 3 | 278 | 134 | |
BC022949.1 | - | 2 - 3 | 255 | 134 | |
BC033103.1 | - | 2 - 3 | 272 | 134 | |
BC111399.1 | - | 2 - 3 | 278 | 134 | |
BC132724.1 | - | 2 - 3 | 302 | 134 | |
BC136599.1 | - | 2 - 3 | 302 | 134 | |
D21064.1 | - | 2 - 3 | 276 | 134 | |
D50913.2 | - | 2 - 3 | 276 | 134 |