The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008779.1 | NP_001008779.1 | 1 - 2 | 255 | 89 |
NM_001142634.1 | NP_001136106.1 | 2 - 3 | 369 | 89 | |
NM_182756.3 | NP_877433.2 | 3 - 4 | 443 | 89 | |
GenBank mRNA | AY820303.1 | - | 1 - 2 | 237 | 89 |
AY820304.1 | - | 1 - 2 | 237 | 89 | |
BC093005.1 | - | 2 - 3 | 369 | 89 |