Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNAT1
Entrez Gene ID
2779
Gene Name
G protein subunit alpha transducin 1
Gene Aliases
CSNB1G, CSNBAD3, GBT1, GNATR
Chromosome Location
Chr.3: 50191610 - 50197696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000172.3 NP_000163.2 5 - 6 694 104
NM_144499.2 NP_653082.1 5 - 6 694 104
GenBank mRNA AF493908.1 - 5 - 6 578 104
BC095505.1 - 5 - 6 581 104
X15088.1 - 5 - 6 681 104

View Full Product Details