The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152834.2 | NP_690047.2 | 2 - 3 | 263 | 80 |
GenBank mRNA | AK125009.1 | - | 1 - 2 | 1148 | 80 |
AL137269.1 | - | 2 - 3 | 263 | 80 | |
BC016010.1 | - | 2 - 3 | 235 | 80 | |
BC032379.1 | - | 2 - 3 | 523 | 80 |