Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM18
Entrez Gene ID
129787
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.2: 667973 - 677439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152834.2 NP_690047.2 4 - 5 421 81
GenBank mRNA AK125009.1 - 3 - 4 1306 81
AL137269.1 - 4 - 5 421 81
BC016010.1 - 4 - 5 393 81
BC032379.1 - 4 - 5 681 81

View Full Product Details