The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319674.1 | NP_001306603.1 | 4 - 5 | 471 | 104 |
NM_006582.3 | NP_006573.2 | 4 - 5 | 501 | 104 | |
NM_024482.2 | NP_077808.1 | 4 - 5 | 468 | 104 | |
XM_011540518.2 | XP_011538820.1 | 4 - 5 | 604 | 104 | |
XM_011540519.2 | XP_011538821.1 | 4 - 5 | 444 | 104 | |
XM_011540521.2 | XP_011538823.1 | 4 - 5 | 573 | 104 | |
XM_011540522.2 | XP_011538824.1 | 4 - 5 | 413 | 104 | |
XM_017000087.1 | XP_016855576.1 | 4 - 5 | 619 | 104 | |
GenBank mRNA | AF099013.1 | - | 3 - 4 | 365 | 104 |
AF173868.1 | - | 4 - 5 | 396 | 104 | |
AF203694.1 | - | 4 - 5 | 395 | 104 | |
AK000892.1 | - | 4 - 5 | 467 | 104 | |
AK023126.1 | - | 4 - 5 | 388 | 104 | |
AK315195.1 | - | 4 - 5 | 447 | 104 | |
AK315436.1 | - | 4 - 5 | 405 | 104 | |
BC001473.2 | - | 4 - 5 | 418 | 104 |