Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALGDS
Entrez Gene ID
5900
Gene Name
ral guanine nucleotide dissociation stimulator
Gene Aliases
RGDS, RGF, RalGEF
Chromosome Location
Chr.9: 133097720 - 133149220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042368.2 NP_001035827.1 12 - 13 1872 97
NM_001271774.1 NP_001258703.1 12 - 13 1784 97
NM_001271775.1 NP_001258704.1 12 - 13 1877 97
NM_001271776.1 NP_001258705.1 12 - 13 1857 97
NM_006266.3 NP_006257.1 12 - 13 1893 97
GenBank mRNA AB037729.1 - 8 - 9 1185 97
AF027169.1 - 12 - 13 1788 97
AF295773.1 - 12 - 13 1808 97
AK000242.1 - 7 - 8 850 97
AK024575.1 - 4 - 5 447 97
AK056462.1 - 11 - 12 1570 97
AK074114.1 - 12 - 13 1763 97
AK090450.1 - 8 - 9 1219 97
AK127524.1 - 12 - 13 1877 97
AK301470.1 - 12 - 13 1744 97
BC033198.1 - 4 - 5 289 97
BC059362.1 - 12 - 13 1847 97

View Full Product Details