The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080606.3 | NP_542173.2 | 1 - 1 | 362 | 84 |
GenBank mRNA | BC137288.1 | - | 1 - 1 | 362 | 84 |
BC137308.1 | - | 1 - 1 | 362 | 84 |