The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007176.4 | NP_001007177.1 | 3 - 4 | 321 | 150 |
NM_001256596.1 | NP_001243525.1 | 3 - 4 | 213 | 150 | |
NM_001256597.1 | NP_001243526.1 | 3 - 4 | 321 | 150 | |
NM_001256598.1 | NP_001243527.1 | 3 - 4 | 321 | 150 | |
GenBank mRNA | AJ276240.1 | - | 3 - 4 | 308 | 150 |
BC017981.1 | - | 3 - 4 | 314 | 150 | |
BG202040.1 | - | 3 - 4 | 239 | 150 | |
DA820911.1 | - | 3 - 4 | 322 | 150 | |
DA900243.1 | - | 3 - 4 | 321 | 150 |