The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144074.2 | NP_001137546.1 | 3 - 4 | 1445 | 99 |
NM_001321594.1 | NP_001308523.1 | 5 - 6 | 1380 | 99 | |
NM_001321596.1 | NP_001308525.1 | 4 - 5 | 1477 | 99 | |
NM_017996.4 | NP_060466.2 | 4 - 5 | 1515 | 99 | |
NR_135739.1 | 4 - 5 | 1306 | 99 | ||
NR_135740.1 | 3 - 4 | 455 | 99 | ||
NR_135741.1 | 4 - 5 | 525 | 99 | ||
NR_135742.1 | 3 - 4 | 1445 | 99 | ||
NR_135743.1 | 6 - 7 | 1408 | 99 | ||
NR_135744.1 | 5 - 6 | 1338 | 99 | ||
GenBank mRNA | AB073887.1 | - | 3 - 4 | 1417 | 99 |
AK000965.1 | - | 2 - 3 | 745 | 99 | |
AK054603.1 | - | 4 - 5 | 1465 | 99 | |
BC001242.2 | - | 2 - 3 | 706 | 99 | |
BC073867.1 | - | 4 - 5 | 1248 | 99 | |
BC109060.1 | - | 3 - 4 | 1304 | 99 | |
BC109061.1 | - | 3 - 4 | 1304 | 99 | |
BG116971.1 | - | 4 - 5 | 455 | 99 | |
BU856989.1 | - | 3 - 4 | 393 | 99 |