Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCH1
Entrez Gene ID
23007
Gene Name
phospholipase C eta 1
Gene Aliases
PLCL3
Chromosome Location
Chr.3: 155458603 - 155745723 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130960.1 NP_001124432.1 9 - 10 1595 71
NM_001130961.1 NP_001124433.1 9 - 10 1595 71
NM_014996.2 NP_055811.1 9 - 10 1338 71
XM_005247238.1 XP_005247295.1 10 - 11 1644 71
XM_005247239.1 XP_005247296.1 9 - 10 1695 71
XM_011512560.2 XP_011510862.1 10 - 11 1653 71
XM_011512561.2 XP_011510863.1 10 - 11 1785 71
XM_011512562.2 XP_011510864.1 11 - 12 2402 71
XM_011512564.2 XP_011510866.1 10 - 11 1644 71
XM_011512565.2 XP_011510867.1 9 - 10 2116 71
XM_011512566.2 XP_011510868.1 9 - 10 1757 71
XM_011512567.1 XP_011510869.1 10 - 11 1644 71
XM_017005923.1 XP_016861412.1 10 - 11 1644 71
XM_017005924.1 XP_016861413.1 10 - 11 1644 71
XM_017005925.1 XP_016861414.1 10 - 11 1644 71
XM_017005926.1 XP_016861415.1 9 - 10 1543 71
XM_017005927.1 XP_016861416.1 10 - 11 1644 71
XM_017005928.1 XP_016861417.1 10 - 11 1644 71
XM_017005929.1 XP_016861418.1 9 - 10 1695 71
GenBank mRNA AB028992.1 - 5 - 6 673 71
AK022610.1 - 1 - 2 147 71
AK096620.1 - 9 - 10 1695 71
AY691170.1 - 9 - 10 1318 71
AY691171.1 - 9 - 10 1318 71
BC113950.1 - 9 - 10 1338 71

View Full Product Details