Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD11
Entrez Gene ID
5717
Gene Name
proteasome 26S subunit, non-ATPase 11
Gene Aliases
Rpn6, S9, p44.5
Chromosome Location
Chr.17: 32444463 - 32483319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270482.1 NP_001257411.1 3 - 4 383 91
NM_002815.3 NP_002806.2 3 - 4 383 91
GenBank mRNA AB003102.1 - 3 - 4 333 91
AK223196.1 - 3 - 4 336 91
AK290602.1 - 3 - 4 372 91
AK300342.1 - 3 - 4 342 91
BC000437.2 - 3 - 4 330 91
BC004430.1 - 3 - 4 330 91
BP246240.1 - 3 - 4 386 91

View Full Product Details