The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014639.3 | NP_055454.1 | 8 - 9 | 837 | 72 |
GenBank mRNA | AB002370.1 | - | 8 - 9 | 837 | 72 |
AK307531.1 | - | 7 - 8 | 665 | 72 | |
BC056893.1 | - | 8 - 9 | 810 | 72 |