The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002876.2 | NP_001002876.1 | 1 - 2 | 148 | 67 |
NM_001304371.1 | NP_001291300.1 | 1 - 2 | 148 | 67 | |
NM_024053.4 | NP_076958.1 | 1 - 2 | 148 | 67 | |
XM_011530368.2 | XP_011528670.1 | 1 - 2 | 135 | 67 | |
GenBank mRNA | AK123479.1 | - | 1 - 2 | 137 | 67 |
AL160131.1 | - | 1 - 2 | 96 | 67 | |
BC000705.2 | - | 1 - 2 | 95 | 67 | |
BC007495.2 | - | 1 - 2 | 108 | 67 | |
BE797487.1 | - | 1 - 2 | 102 | 67 | |
BQ922403.1 | - | 1 - 2 | 81 | 67 | |
BU556693.1 | - | 1 - 2 | 148 | 67 | |
CA414668.1 | - | 3 - 4 | 67 | ||
CR456399.1 | - | 1 - 2 | 121 | 67 | |
CR457392.1 | - | 1 - 2 | 60 | 67 |