The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258461.1 | NP_001245390.1 | 5 - 6 | 1078 | 84 |
NM_006712.4 | NP_006703.1 | 5 - 6 | 1159 | 84 | |
NM_033015.3 | NP_148936.2 | 4 - 5 | 736 | 84 | |
XM_005249932.1 | XP_005249989.1 | 5 - 6 | 1351 | 84 | |
XM_005249933.3 | XP_005249990.1 | 5 - 6 | 1557 | 84 | |
XM_011515761.2 | XP_011514063.1 | 5 - 6 | 1159 | 84 | |
XM_011515762.2 | XP_011514064.1 | 5 - 6 | 1159 | 84 | |
XM_011515763.1 | XP_011514065.1 | 4 - 5 | 617 | 84 | |
XM_017011704.1 | XP_016867193.1 | 5 - 6 | 1270 | 84 | |
XM_017011705.1 | XP_016867194.1 | 4 - 5 | 714 | 84 | |
XM_017011706.1 | XP_016867195.1 | 4 - 5 | 906 | 84 | |
GenBank mRNA | AB209783.1 | - | 5 - 6 | 979 | 84 |
AK223132.1 | - | 5 - 6 | 1069 | 84 | |
AK292232.1 | - | 5 - 6 | 1044 | 84 | |
AL713798.1 | - | 3 - 4 | 2012 | 84 | |
BC000377.2 | - | 5 - 6 | 1071 | 84 | |
BC006386.2 | - | 4 - 5 | 1042 | 84 | |
BC011770.2 | - | 5 - 6 | 1070 | 84 | |
BC039026.2 | - | 4 - 5 | 706 | 84 | |
BM799420.1 | - | 2 - 3 | 407 | 84 | |
X86779.1 | - | 5 - 6 | 1063 | 84 |