The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004120.4 | NP_004111.2 | 3 - 4 | 623 | 77 |
GenBank mRNA | AK057968.1 | - | 3 - 4 | 536 | 77 |
AK226018.1 | - | 3 - 4 | 549 | 77 | |
AK314325.1 | - | 3 - 4 | 578 | 77 | |
BC022272.1 | - | 3 - 4 | 578 | 77 | |
BC073163.1 | - | 3 - 4 | 560 | 77 | |
CR457062.1 | - | 2 - 3 | 309 | 77 | |
M55543.1 | - | 3 - 4 | 465 | 77 |