The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000810.3 | NP_000801.1 | 2 - 3 | 456 | 78 |
NM_001165037.1 | NP_001158509.1 | 2 - 3 | 322 | 78 | |
XM_005268258.2 | XP_005268315.1 | 2 - 3 | 334 | 78 | |
XM_006720459.2 | XP_006720522.1 | 2 - 3 | 238 | 78 | |
XM_017022055.1 | XP_016877544.1 | 2 - 3 | 204 | 78 | |
XM_017022056.1 | XP_016877545.1 | 2 - 3 | 238 | 78 | |
GenBank mRNA | A28104.1 | - | 2 - 3 | 221 | 78 |
AK290453.1 | - | 2 - 3 | 119 | 78 | |
BC111979.1 | - | 1 - 2 | 68 | 78 | |
BC113422.1 | - | 1 - 2 | 68 | 78 | |
DA157081.1 | - | 2 - 3 | 456 | 78 | |
DB506215.1 | - | 2 - 3 | 324 | 78 | |
L08485.1 | - | 2 - 3 | 229 | 78 |