The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000810.3 | NP_000801.1 | 7 - 8 | 1114 | 80 |
NM_001165037.1 | NP_001158509.1 | 7 - 8 | 980 | 80 | |
XM_005268258.2 | XP_005268315.1 | 7 - 8 | 992 | 80 | |
XM_006720459.2 | XP_006720522.1 | 7 - 8 | 896 | 80 | |
XM_017022055.1 | XP_016877544.1 | 7 - 8 | 862 | 80 | |
XM_017022056.1 | XP_016877545.1 | 7 - 8 | 896 | 80 | |
GenBank mRNA | A28104.1 | - | 7 - 8 | 879 | 80 |
AK290453.1 | - | 7 - 8 | 777 | 80 | |
AK294303.1 | - | 4 - 5 | 437 | 80 | |
AK303742.1 | - | 2 - 3 | 285 | 80 | |
BC111979.1 | - | 6 - 7 | 726 | 80 | |
BC113422.1 | - | 6 - 7 | 726 | 80 | |
BT009830.1 | - | 5 - 6 | 582 | 80 | |
L08485.1 | - | 7 - 8 | 887 | 80 |