The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144825.1 | NP_001138297.1 | 3 - 4 | 647 | 66 |
NM_001144826.1 | NP_001138298.1 | 3 - 4 | 632 | 66 | |
NM_006695.4 | NP_006686.1 | 3 - 4 | 647 | 66 | |
XM_005256964.3 | XP_005257021.1 | 3 - 4 | 580 | 66 | |
XM_017024035.1 | XP_016879524.1 | 3 - 4 | 647 | 66 | |
XM_017024036.1 | XP_016879525.1 | 3 - 4 | 568 | 66 | |
XM_017024037.1 | XP_016879526.1 | 3 - 4 | 580 | 66 | |
XM_017024038.1 | XP_016879527.1 | 3 - 4 | 647 | 66 | |
XM_017024039.1 | XP_016879528.1 | 3 - 4 | 647 | 66 | |
XM_017024040.1 | XP_016879529.1 | 3 - 4 | 568 | 66 | |
GenBank mRNA | AB209802.1 | - | 3 - 4 | 631 | 66 |
AF055026.1 | - | 3 - 4 | 411 | 66 | |
AK123330.1 | - | 3 - 4 | 1061 | 66 | |
AK226045.1 | - | 3 - 4 | 608 | 66 | |
AK313669.1 | - | 3 - 4 | 568 | 66 | |
AL133657.1 | - | 3 - 4 | 793 | 66 | |
BC006194.2 | - | 3 - 4 | 610 | 66 | |
BX537443.1 | - | 3 - 4 | 529 | 66 | |
CR456748.1 | - | 3 - 4 | 373 | 66 | |
U93871.1 | - | 3 - 4 | 358 | 66 |