The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318970.1 | NP_001305899.1 | 3 - 4 | 213 | 106 |
NM_001623.4 | NP_001614.3 | 3 - 4 | 274 | 106 | |
XM_005248870.4 | XP_005248927.1 | 3 - 4 | 266 | 106 | |
XM_017010332.1 | XP_016865821.1 | 3 - 4 | 210 | 106 | |
GenBank mRNA | AF088038.1 | - | 3 - 4 | 220 | 106 |
AK290771.1 | - | 3 - 4 | 209 | 106 | |
AV716806.1 | - | 3 - 4 | 194 | 106 | |
AV735626.1 | - | 3 - 4 | 320 | 106 | |
BC009474.1 | - | 3 - 4 | 253 | 106 | |
CD521003.1 | - | 3 - 4 | 181 | 106 | |
D86438.1 | - | 3 - 4 | 162 | 106 | |
EF070983.1 | - | 3 - 4 | 153 | 106 | |
JQ782414.1 | - | 3 - 4 | 248 | 106 | |
JQ782415.1 | - | 3 - 4 | 187 | 106 | |
U19713.1 | - | 3 - 4 | 177 | 106 |