Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLIM4
Entrez Gene ID
27333
Gene Name
golgi integral membrane protein 4
Gene Aliases
GIMPC, GOLPH4, GPP130, P138
Chromosome Location
Chr.3: 168008672 - 168095925 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308155.1 NP_001295084.1 4 - 5 1010 72
NM_014498.4 NP_055313.1 4 - 5 1010 72
XM_005247364.2 XP_005247421.1 4 - 5 1009 72
XM_005247365.2 XP_005247422.1 4 - 5 1009 72
XM_017006183.1 XP_016861672.1 4 - 5 489 72
GenBank mRNA AK307557.1 - 4 - 5 715 72
AK307718.1 - 4 - 5 685 72
U55853.1 - 4 - 5 383 72

View Full Product Details