The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293273.1 | NP_001280202.1 | 6 - 7 | 763 | 125 |
NM_020166.4 | NP_064551.3 | 8 - 9 | 1022 | 125 | |
NR_120639.1 | 6 - 7 | 789 | 125 | ||
NR_120640.1 | 8 - 9 | 1542 | 125 | ||
XM_006713702.1 | XP_006713765.1 | 7 - 8 | 828 | 125 | |
XM_011512992.2 | XP_011511294.1 | 8 - 9 | 1620 | 125 | |
XM_011512993.1 | XP_011511295.1 | 8 - 9 | 1022 | 125 | |
XM_017006874.1 | XP_016862363.1 | 7 - 8 | 1387 | 125 | |
XM_017006875.1 | XP_016862364.1 | 6 - 7 | 624 | 125 | |
GenBank mRNA | AB029826.1 | - | 8 - 9 | 1007 | 125 |
AB209737.1 | - | 8 - 9 | 923 | 125 | |
AF297332.1 | - | 8 - 9 | 1041 | 125 | |
AF310339.1 | - | 8 - 9 | 1007 | 125 | |
AF310972.1 | - | 8 - 9 | 1008 | 125 | |
AK023051.1 | - | 8 - 9 | 998 | 125 | |
AK294882.1 | - | 6 - 7 | 720 | 125 | |
AK295293.1 | - | 7 - 8 | 844 | 125 | |
AK316428.1 | - | 7 - 8 | 773 | 126 | |
AL442091.1 | - | 8 - 9 | 941 | 125 | |
BC004187.2 | - | 8 - 9 | 1007 | 125 | |
BC004214.2 | - | 8 - 9 | 1007 | 125 | |
BC036395.1 | - | 7 - 8 | 1256 | 125 | |
KU178727.1 | - | 8 - 9 | 875 | 125 |