Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNE2
Entrez Gene ID
9992
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Gene Aliases
ATFB4, LQT5, LQT6, MIRP1
Chromosome Location
Chr.21: 34364024 - 34371141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172201.1 NP_751951.1 1 - 2 134 123
GenBank mRNA AF302095.1 - 1 - 2 134 123
BC093892.1 - 1 - 2 85 123
BC112087.1 - 1 - 2 86 123
DQ192290.1 - 1 - 2 134 123

View Full Product Details