Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MFSD11
- Entrez Gene ID
-
79157
- Gene Name
- major facilitator superfamily domain containing 11
- Gene Aliases
-
ET
- Chromosome Location
-
Chr.17: 76736565 - 76803805 on Build GRCh38
View Full Product Details